B20169.06
B20169.14
B20169.22
B20169.06EA
21.7
EUR
InStock
B20169.06
B20169.14
B20169.22
Methyl-2-chlor-4-nitrobenzoat ≥98%
Methyl-2-chlor-4-nitrobenzoat
Purity: 98%
CAS No.: 13324-11-3
Molecular Formula: C8H6ClNO4
Formula Weight: 215.59
Melting Point: 74-78°
Beilstein Reference: 1877723