B20233.18
B20233.22
B20233.30
B20233.18EA
46
EUR
InStock
B20233.18
B20233.22
B20233.30
Methyl-N-methylanthranilat ≥98%
Methyl-N-methylanthranilat
Purity: 98%
CAS No.: 85-91-6
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Flash Point: 133°(271°F)
Refractive Index: 1.5800
Melting Point: 17-19°
Boiling Point: 256-258°
Beilstein Reference: 607217
MDL No.: MFCD00017183