B20379.03
B20379.06
B20379.03EA
36
EUR
InStock
B20379.03
B20379.06
2,3,6-Trifluorphenol ≥98%
2,3,6-Trifluorphenol
Purity: 98%
CAS No.: 113798-74-6
Molecular Formula: C6H3F3O
Formula Weight: 148.09
Flash Point: 63°(145°F)
Melting Point: 31-36°
Beilstein Reference: 4976927
UN No.: 3261
MDL No.: MFCD00061214