B20567.06
B20567.14
B20567.22
B20567.06EA
46.6
EUR
InStock
B20567.06
B20567.14
B20567.22
3-Amino-5-tert-butylisoxazol 97%
3-Amino-5-tert-butylisoxazol
Purity: 97%
CAS No.: 55809-36-4
Molecular Formula: C7H12N2O
Formula Weight: 140.19
Melting Point: 110-113°
Beilstein Reference: 1100990