B21000.14
B21000.06
B21000.22
B21000.14EA
117
EUR
InStock
B21000.14
B21000.06
B21000.22
Cyclohexylmethylketon 95%
Cyclohexylmethylketon
Purity: 95%
CAS No.: 823-76-7
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 66°(151°F)
Physical Form: Liquid
Density: 0.917
Refractive Index: 1.4520
Melting Point: -34°
Boiling Point: 181-183°
Beilstein Reference: 507229
MDL No.: MFCD00040418