B21127.06
B21127.14
B21127.22
B21127.06EA
51.2
EUR
InStock
B21127.06
B21127.14
B21127.22
Triisopropylsilyltrifluormethansulfonat ≥97%
Triisopropylsilyltrifluormethansulfonat
Purity: 97%
CAS No.: 80522-42-5
Molecular Formula: CF3SO3Si[CH(CH3)2]3
Formula Weight: 306.42
Flash Point: 100°(212°F)
Refractive Index: 1.4150
Boiling Point: 45-46°/0.03mm
Beilstein Reference: 3591541
UN No.: 3265
MDL No.: MFCD00009913