B21176.22
B21176.36
B21176.22EA
54.1
EUR
InStock
B21176.22
B21176.36
Vinyltrimethoxysilan ≥98%
Vinyltrimethoxysilan
Purity: 98%
CAS No.: 2768-02-7
Molecular Formula: C5H12O3Si
Formula Weight: 148.24
Flash Point: 23°(73°F)
Refractive Index: 1.3920
Odour: Fruit-like
Melting Point: -40°
Boiling Point: 122-124°
Beilstein Reference: 1099136
UN No.: 1993
MDL No.: MFCD00008605