B21191.14
B21191.22
B21191.14EA
81.9
EUR
InStock
B21191.14
B21191.22
(3-Mercaptopropyl)triethoxysilan ≥94%
(3-Mercaptopropyl)triethoxysilan
Purity: 94%
CAS No.: 14814-09-6
Molecular Formula: C9H22O3SSi
Formula Weight: 238.42
Flash Point: 88°(190°F)
Physical Form: Liquid
Density: 0.98
Refractive Index: 1.4370
Sensitivity: Air and Moisture Sensitive
Appearance: Colourless
Odour: Unpleasant
Boiling Point: 210°
Beilstein Reference: 2039575
MDL No.: MFCD00053924