B21267.06
B21267.14
B21267.06EA
39.9
EUR
InStock
B21267.06
B21267.14
2-Methyl-3-nitrobenzotrifluorid ≥97%
2-Methyl-3-nitrobenzotrifluorid
Purity: 97%
CAS No.: 6656-49-1
Molecular Formula: C8H6F3NO2
Formula Weight: 205.14
Storage Temperature: 2°C to 8°C
Flash Point: 100°(212°F)
Refractive Index: 1.4780
Boiling Point: 86°/10mm
Beilstein Reference: 2457216
UN No.: 2810
MDL No.: MFCD00042322