B21352.03
B21352.06
B21352.14
B21352.03EA
33.1
EUR
InStock
B21352.03
B21352.06
B21352.14
4-Fluorphthalsäureanhydrid ≥98%
4-Fluorphthalsäureanhydrid
Purity: 98%
CAS No.: 319-03-9
Molecular Formula: C8H3FO3
Formula Weight: 166.11
Melting Point: 74-79°
Boiling Point: 147-149°/20mm
MDL No.: MFCD00191363