B21400.06
B21400.14
B21400.06EA
56.8
EUR
InStock
B21400.06
B21400.14
3,5-Difluorbenzoylchlorid ≥98%
3,5-Difluorbenzoylchlorid
Purity: 98+%
CAS No.: 129714-97-2
Molecular Formula: C7H3ClF2O
Formula Weight: 176.55
Flash Point: 69°(156°F)
Refractive Index: 1.5030
Boiling Point: 173-175°
Beilstein Reference: 4307685
UN No.: 3265
MDL No.: MFCD00010309