B21408.14
B21408.22
B21408.14EA
46.6
EUR
InStock
B21408.14
B21408.22
Tetracyclin Hydrochlorid 96%
Tetracyclin Hydrochlorid
Purity: 96%
CAS No.: 64-75-5
Molecular Formula: C22H24N2O8.HCl
Formula Weight: 480.91
Melting Point: 220-223°
Merck Reference: 14,9196
Optical Rotation: -247° (c=1, 0.1M HCl)
MDL No.: MFCD00078142
Can induce apoptosis in osteoclasts