B21586.AD
B21586.AK
B21586.AU
B21586.ADEA
24.1
EUR
InStock
B21586.AD
B21586.AK
B21586.AU
2-Chlorbutan ≥98%
2-Chlorbutan
Purity: 98+%
CAS No.: 78-86-4
Molecular Formula: CH3CHClCH2CH3
Formula Weight: 92.57
Flash Point: -15°(5°F)
Physical Form: Liquid
Density: 0.873
Refractive Index: 1.3970
Odour: Sweet
Melting Point: -141 to -140°
Boiling Point: 67-69°
Beilstein Reference: 1718770
Merck Reference: 14,1561
UN No.: 1127
MDL No.: MFCD00000871