B21595.03
B21595.06
B21595.14
B21595.03EA
30.2
EUR
InStock
B21595.03
B21595.06
B21595.14
(2,5-Difluorphenyl)essigsäure ≥98%
(2,5-Difluorphenyl)essigsäure
Purity: 98%
CAS No.: 85068-27-5
Molecular Formula: C8H6F2O2
Formula Weight: 172.13
Melting Point: 123-127°
Beilstein Reference: 6844956