B21819.09
B21819.18
B21819.30
B21819.09EA
34.2
EUR
InStock
B21819.09
B21819.18
B21819.30
TES (2-(Tris(hydroxymethyl)methylamino)ethansulfonsäure) ≥99%
TES (2-(Tris(hydroxymethyl)methylamino)ethansulfonsäure)
Purity: 99%
CAS No.: 7365-44-8
Molecular Formula: C6H15NO6S
Formula Weight: 229.25
Odour: Odourless
Melting Point: ca 225° dec.
Beilstein Reference: 1957061
Merck Reference: 14,9176
MDL No.: MFCD00007532