B21825.0F
B21825.AE
B21825.AP
B21825.0FEA
150
EUR
InStock
B21825.0F
B21825.AE
B21825.AP
(±)-2-Methyl-1-butanol ≥98%
(±)-2-Methyl-1-butanol
Purity: 98%
CAS No.: 137-32-6
Molecular Formula: C5H12O
Formula Weight: 88.15
Flash Point: 43°(109°F)
Refractive Index: 1.4110
Melting Point: -70°
Boiling Point: 129-131°
Beilstein Reference: 1718810
Merck Reference: 14,6030
UN No.: 1105
MDL No.: MFCD00004743