B21964.22
B21964.36
B21964.22EA
22
EUR
InStock
B21964.22
B21964.36
4-Chlor-3,5-xylenol ≥98%
4-Chlor-3,5-xylenol
Purity: 98+%
CAS No.: 88-04-0
Molecular Formula: C8H9ClO
Formula Weight: 156.61
Flash Point: 138°(280°F)
Odour: Odourless
Boiling Point: ca 246°
Beilstein Reference: 1862539
Merck Reference: 14,2176
MDL No.: MFCD00002324