B22085.14
B22085.22
B22085.36
B22085.14EA
32.3
EUR
InStock
B22085.14
B22085.22
B22085.36
Nickel(II)chlorid, wasserfrei 98%
Nickel(II)chlorid
Purity: 98%
CAS No.: 7718-54-9
Molecular Formula: Cl2Ni
Formula Weight: 129.60
Odour: Odourless
Melting Point: 1001°
UN No.: 3288
MDL No.: MFCD00011142