B22098.06
B22098.14
B22098.06EA
63.3
EUR
InStock
B22098.06
B22098.14
4-Amino-2-chlorpyridin 98%
4-Amino-2-chlorpyridin
Purity: 98%
CAS No.: 14432-12-3
Molecular Formula: C5H5ClN2
Formula Weight: 128.56
Odour: Unpleasant
Melting Point: 91-94°
Boiling Point: 153°/5mm
Beilstein Reference: 108671
MDL No.: MFCD00060089