B22157.06
B22157.14
B22157.22
B22157.06EA
43
EUR
InStock
B22157.06
B22157.14
B22157.22
4-Cyanphenylhydrazin Hydrochlorid ≥97%
4-Cyanphenylhydrazin Hydrochlorid
Purity: 97%
CAS No.: 2863-98-1
Molecular Formula: C7H7N3.HCl
Formula Weight: 169.61
Melting Point: ca 243° dec.
Beilstein Reference: 3565486