B22171.06
B22171.14
B22171.06EA
56.7
EUR
InStock
B22171.06
B22171.14
2-Furylacetone 99%
2-Furylacetone
Purity: 99%
CAS No.: 6975-60-6
Molecular Formula: C7H8O2
Formula Weight: 124.14
Storage Temperature: 2°C to 8°C
Flash Point: 65°(149°F)
Odour: Stench
Boiling Point: 179-180°
MDL No.: MFCD02683084