B22185.03
B22185.06
B22185.14
B22185.03EA
48.3
EUR
InStock
B22185.03
B22185.06
B22185.14
1-Brom-3-chlor-4-iodbenzol ≥97%
4-Bromo-2-chloro-1-iodobenzene
Purity: 97%
CAS No.: 31928-47-9
Molecular Formula: C6H3BrClI
Formula Weight: 317.36
Refractive Index: 1.6722
Melting Point: 33-35°
Boiling Point: 103-105°/1mm
Beilstein Reference: 2435075
MDL No.: MFCD00079709