B22206.06
B22206.06EA
34.4
EUR
InStock
B22206.06
4-(Methylthio)butanon ≥98%
4-(Methylthio)butanon
Purity: 98%
CAS No.: 34047-39-7
Molecular Formula: CH3SCH2CH2COCH3
Formula Weight: 118.20
Flash Point: 70°(158°F)
Refractive Index: 1.473
Boiling Point: 105-107°/55mm
MDL No.: MFCD00040029