B22223.06
B22223.14
B22223.22
B22223.06EA
32.9
EUR
InStock
B22223.06
B22223.14
B22223.22
2,3-Dimethylbutan ≥99%
2,3-Dimethylbutane
Purity: 99%
CAS No.: 79-29-8
Molecular Formula: C6H14
Formula Weight: 86.18
Flash Point: -29°(-20°F)
Refractive Index: 1.375
Odour: Mild
Melting Point: -129°
Boiling Point: 57-59°
Beilstein Reference: 1730737
UN No.: 2457
MDL No.: MFCD00008925