B22443.03
B22443.06
B22443.14
B22443.03EA
41
EUR
InStock
B22443.03
B22443.06
B22443.14
2-(Trifluormethyl)acrylsäure ≥98%
2-(Trifluormethyl)acrylsäure
Purity: 98%
CAS No.: 381-98-6
Molecular Formula: C4H3F3O2
Formula Weight: 140.06
Flash Point: 82°(180°F)
Melting Point: 49-51°
UN No.: 3261
MDL No.: MFCD00042424