B22525.06
B22525.14
B22525.06EA
21.4
EUR
InStock
B22525.06
B22525.14
Ethyl-2-bromoisovalerate 97%
Ethyl-2-bromoisovalerate
Purity: 97%
CAS No.: 609-12-1
Molecular Formula: C7H13BrO2
Formula Weight: 209.09
Flash Point: 65°(149°F)
Refractive Index: 1.4490
Odour: Pungent
Boiling Point: 185-186°
Beilstein Reference: 1099039
UN No.: UN3265
MDL No.: MFCD00026855