B22556.09
B22556.18
B22556.09EA
33.3
EUR
InStock
B22556.09
B22556.18
2-Mercapto-1,3,4-thiadiazol ≥98%
2-Mercapto-1,3,4-thiadiazole
Purity: 98%
CAS No.: 18686-82-3
Molecular Formula: C2H2N2S2
Formula Weight: 118.17
Melting Point: 142-144°
MDL No.: MFCD00040404