B22697.14
B22697.22
B22697.36
B22697.14EA
39.3
EUR
InStock
B22697.14
B22697.22
B22697.36
2-Iodtoluol ≥98%
2-Iodtoluol
Purity: 98%
CAS No.: 615-37-2
Molecular Formula: C7H7I
Formula Weight: 218.04
Flash Point: 90°(194°F)
Refractive Index: 1.6090
Boiling Point: 210-212°
Beilstein Reference: 1634294
MDL No.: MFCD00001042