B22705.14
B22705.03
B22705.06
B22705.14EA
308
EUR
InStock
B22705.14
B22705.03
B22705.06
2-Amino-5-iodpyrimidin 97%
2-Amino-5-iodopyrimidine
Purity: 97%
CAS No.: 1445-39-2
Molecular Formula: C4H4IN3
Formula Weight: 221.00
Melting Point: 217-220°
MDL No.: MFCD01075666