B22781.03
B22781.06
B22781.14
B22781.03EA
35.2
EUR
InStock
B22781.03
B22781.06
B22781.14
2,3-Dichlorphenylboronsäure ≥98%
2,3-Dichlorphenylboronsäure
Purity: 98%
CAS No.: 151169-74-3
Molecular Formula: C6H5BCl2O2
Formula Weight: 190.82
Melting Point: 233-242°
MDL No.: MFCD01075703