B22805.03
B22805.06
B22805.14
B22805.03EA
20.7
EUR
InStock
B22805.03
B22805.06
B22805.14
2,6-Difluorphenylboronsäure ≥98%
2,6-Difluorphenylboronsäure
Purity: 98%
CAS No.: 162101-25-9
Molecular Formula: C6H5BF2O2
Formula Weight: 157.91
Melting Point: ca 140° dec.
Beilstein Reference: 8545931