B23028.14
B23028.22
B23028.36
B23028.14EA
28.5
EUR
InStock
B23028.14
B23028.22
B23028.36
Hexamethylcyclotrisiloxan ≥97%
Hexamethylcyclotrisiloxane
Purity: 97%
CAS No.: 541-05-9
Molecular Formula: C6H18O3Si3
Formula Weight: 222.47
Flash Point: 35°(95°F)
Melting Point: 64-66°
Boiling Point: 134°
Beilstein Reference: 1766829
UN No.: 1325
MDL No.: MFCD00005943