B23037.06
B23037.06EA
104
EUR
InStock
B23037.06
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 96%
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
Purity: 96%
CAS No.: 14347-78-5
Molecular Formula: C6H12O3
Formula Weight: 132.18
Storage Temperature: 2°C to 8°C
Flash Point: 80°(176°F)
Refractive Index: 1.4350
Boiling Point: 82-83°/14mm
Beilstein Reference: 80117
Merck Reference: 14,5213
Optical Rotation: -11.5° (c=5 in methanol)
MDL No.: MFCD00003213