B23173.06
B23173.14
B23173.22
B23173.06EA
62.9
EUR
InStock
B23173.06
B23173.14
B23173.22
Tetraethylammoniumfluorid Hydrat ≥97%
Tetraethylammoniumfluorid Hydrat
Purity: 97%
CAS No.: 98330-04-2
Molecular Formula: (CH3CH2)4NF.xH2O
Formula Weight: 149.26(anhy)
Beilstein Reference: 4152659
MDL No.: MFCD00149990