B23254.14
B23254.22
B23254.14EA
47.8
EUR
InStock
B23254.14
B23254.22
(3-Brompropyl)ammoniumbromid 98%
3-Brompropylamine hydrobromide
Purity: 98%
CAS No.: 5003-71-4
Molecular Formula: Br(CH2)3NH2.HBr
Formula Weight: 218.93
Melting Point: 172-174°
Beilstein Reference: 3906418