B23423.18
B23423.30
B23423.18EA
28.9
EUR
InStock
B23423.18
B23423.30
1-Chlor-3-nitrobenzol ≥98%
1-Chlor-3-nitrobenzol
Purity: 98%
CAS No.: 121-73-3
Molecular Formula: C6H4ClNO2
Formula Weight: 157.56
Flash Point: 124°(255°F)
Melting Point: 43-47°
Boiling Point: 235-236°
Beilstein Reference: 1907947
Merck Reference: 14,2151
UN No.: 1578
MDL No.: MFCD00007202