B23551.06
B23551.14
B23551.22
B23551.06EA
33.3
EUR
InStock
B23551.06
B23551.14
B23551.22
Chlorbutyldimethylsilan 96%
Chlorbutyldimethylsilan
Purity: 96%
CAS No.: 1000-50-6
Molecular Formula: CH3(CH2)3Si(CH3)2Cl
Formula Weight: 150.73
Flash Point: 27°(80°F)
Refractive Index: 1.4200
Boiling Point: 138°
Beilstein Reference: 1732573
UN No.: 2920
MDL No.: MFCD00015727