B23745.06
B23745.14
B23745.06EA
68.6
EUR
InStock
B23745.06
B23745.14
L(+)-Leucinol ≥97%
L(+)-Leucinol
Purity: 97%
CAS No.: 7533-40-6
Molecular Formula: C6H15NO
Formula Weight: 117.19
Flash Point: 90°(194°F)
Refractive Index: 1.4510
Boiling Point: 198-200°
Beilstein Reference: 1719240
Optical Rotation: +1.1° (neat)
MDL No.: MFCD00063676