Stabilization:
0.10 % Stabilisiert mit 4-tert-Butylbrenzcatechin (TBC)
B23806.06
B23806.14
B23806.22
B23806.06EA
75.3
EUR
InStock
B23806.06
B23806.14
B23806.22
3-Chlorstyrol ≥98% stabilisiert
3-Chlorstyrol
Stabilized with 0.1% 4-tert-butylcatechol
Purity: 98%
CAS No.: 2039-85-2
Molecular Formula: C8H7Cl
Formula Weight: 138.60
Storage Temperature: 2°C to 8°C
Flash Point: 67°(153°F)
Refractive Index: 1.5620
Odour: Characteristic
Boiling Point: 188-189°
Beilstein Reference: 2038489
MDL No.: MFCD00000598