B23858.14
B23858.22
B23858.14EA
40
EUR
InStock
B23858.14
B23858.22
2-Phenylimidazol ≥98%
2-Phenylimidazol
Purity: 98+%
CAS No.: 670-96-2
Molecular Formula: C9H8N2
Formula Weight: 144.18
Flash Point: 200°(392°F)
Melting Point: 146-150°
Boiling Point: 198-200°/7mm
Beilstein Reference: 114011
MDL No.: MFCD00005186