B23979.14
B23979.22
B23979.14EA
47.9
EUR
InStock
B23979.14
B23979.22
1-Brom-4-butylbenzol ≥98%
1-Brom-4-butylbenzol
Purity: 98%
CAS No.: 41492-05-1
Molecular Formula: C10H13Br
Formula Weight: 213.12
Flash Point: 108°(226°F)
Refractive Index: 1.5300
Boiling Point: 242°
Beilstein Reference: 2080263
MDL No.: MFCD00040934