B24062.06
B24062.14
B24062.06EA
50.8
EUR
InStock
B24062.06
B24062.14
4-Amino-2,2,6,6-tetramethylpiperidin 98%
4-Amino-2,2,6,6-tetramethylpiperidin
Purity: 98%
CAS No.: 36768-62-4
Molecular Formula: C9H20N2
Formula Weight: 156.27
Flash Point: 75°(167°F)
Melting Point: 16-18°
Boiling Point: 188-189°
MDL No.: MFCD00005984