B24066.03
B24066.06
B24066.14
B24066.03EA
41.1
EUR
InStock
B24066.03
B24066.06
B24066.14
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazin-4,4''-disulfonsäure Mononatriumsalz Hydrat ≥97%
Ferrozine mono-sodium salt hydrate
Purity: 97%
CAS No.: 69898-45-9
Molecular Formula: C20H13N4NaO6S2.xH2O
Formula Weight: 492.47(anhy)
Beilstein Reference: 5222087
MDL No.: MFCD00150794