B24095.AK
B24095.AU
B24095.AKEA
31.9
EUR
InStock
B24095.AK
B24095.AU
1-Amino-3-methoxypropan 99%
1-Amino-3-methoxypropane
Purity: 99%
CAS No.: 5332-73-0
Molecular Formula: CH3O(CH2)3NH2
Formula Weight: 89.14
Flash Point: 27°(81°F)
Refractive Index: 1.4175
Odour: Amine-like
Melting Point: -70°
Boiling Point: 117-119°
Beilstein Reference: 878144
UN No.: 2734
MDL No.: MFCD00014831