B24143.06
B24143.14
B24143.06EA
46.4
EUR
InStock
B24143.06
B24143.14
3-(Trifluormethyl)benzylamin 97%
3-(Trifluormethyl)benzylamin
Purity: 97%
CAS No.: 2740-83-2
Molecular Formula: C8H8F3N
Formula Weight: 175.15
Flash Point: 71°(159°F)
Refractive Index: 1.4630
Boiling Point: 93-97°/28mm
Beilstein Reference: 1102465
UN No.: 2735
MDL No.: MFCD00008117