B24177.06
B24177.14
B24177.22
B24177.06EA
30.3
EUR
InStock
B24177.06
B24177.14
B24177.22
Ethyliodoacetat ≥98%
Ethyliodoacetat
Purity: 98%
CAS No.: 623-48-3
Molecular Formula: ICH2CO2CH2CH3
Formula Weight: 214
Storage Temperature: 2°C to 8°C
Flash Point: 76°(169°F)
Refractive Index: 1.503
Boiling Point: 179-180°
Beilstein Reference: 741934
UN No.: 2927
MDL No.: MFCD00001081