B24232.03
B24232.06
B24232.03EA
55.3
EUR
InStock
B24232.03
B24232.06
2,2-Difluor-1,3-benzodioxol-5-carbaldehyd ≥97%
2,2-Difluor-1,3-benzodioxol-5-carbaldehyd
Purity: 97%
CAS No.: 656-42-8
Molecular Formula: C8H4F2O3
Formula Weight: 186.11
Refractive Index: 1.4990
Boiling Point: 103-105°/20mm
MDL No.: MFCD00792420