B24310.14
B24310.22
B24310.36
B24310.14EA
47.7
EUR
InStock
B24310.14
B24310.22
B24310.36
Kongorot Indikator
Kongorot
CAS No.: 573-58-0
Molecular Formula: C32H22N6Na2O6S2
Formula Weight: 696.67
Odour: Odourless
Melting Point: 360°
Beilstein Reference: 3894858
Merck Reference: 14,2498
MDL No.: MFCD00004028