B24329.06
B24329.14
B24329.06EA
52.8
EUR
InStock
B24329.06
B24329.14
3-Methoxyphenylisocyanat ≥99%
3-Methoxyphenylisocyanat
Purity: 99%
CAS No.: 18908-07-1
Molecular Formula: C8H7NO2
Formula Weight: 149.15
Flash Point: 96°(204°F)
Refractive Index: 1.5430
Odour: Pungent
Boiling Point: 94-95°/10mm
Beilstein Reference: 775986
UN No.: 2206
MDL No.: MFCD00002019