B24374.03
B24374.06
B24374.14
B24374.03EA
46.3
EUR
InStock
B24374.03
B24374.06
B24374.14
2,4-Dimethoxyphenylboronsäure ≥98%
2,4-Dimethoxyphenylboronsäure
Purity: 98%
CAS No.: 133730-34-4
Molecular Formula: C8H11BO4
Formula Weight: 181.98
Melting Point: 115-120°
Beilstein Reference: 4744213